MilliporeSigma
  • Determination of the Absolute Configuration of β-Chiral Primary Alcohols Using the Competing Enantioselective Conversion Method.
Determination of the Absolute Configuration of β-Chiral Primary Alcohols Using the Competing Enantioselective Conversion Method.
Organic letters (2017-05-17)
Alexander S Burns, Alexander J Wagner, Jennifer L Fulton, Kyle Young, Armen Zakarian, Scott D Rychnovsky
ABSTRACT
A method for determining the absolute configuration of β-chiral primary alcohols has been developed. Enantioenriched alcohols were acylated in the presence of either enantiomer of the enantioselective acylation catalyst HBTM, and the faster reaction was determined by measuring product conversion using
Sigma-Aldrich
(2R)-2-Phenyl-3,4-dihydro-2H-pyrimido[2,1-b][1,3]benzothiazole, ≥95%