MilliporeSigma
  • A combined approach for enhancing the stability of recombinant cis-dihydrodiol naphthalene dehydrogenase from Pseudomonas putida G7 allowed for the structural and kinetic characterization of the enzyme.
A combined approach for enhancing the stability of recombinant cis-dihydrodiol naphthalene dehydrogenase from Pseudomonas putida G7 allowed for the structural and kinetic characterization of the enzyme.
Protein expression and purification (2017-01-17)
Débora Maria Abrantes Costa, Mariana Amalia Figueiredo Costa, Samuel Leite Guimarães, Juliana Barbosa Coitinho, Stefanya Velásquez Gómez, Tiago Antônio da Silva Brandão, Ronaldo Alves Pinto Nagem
ABSTRACT
The second enzyme of the naphthalene degradation pathway in Pseudomonas putida G7 is NahB, a dehydrogenase that converts cis-1,2-dihydroxy-1,2-dihydronaphthalene to 1,2-dihydroxynaphthalene. We report the cloning, optimization of expression, purification, kinetic studies and preliminary structural characterization of the recombinant NahB. The nahB gene was cloned into a T7 expression vector and the enzyme was overexpressed in Escherichia coli Rosetta (DE3) as an N-terminal hexa-histidine-tagged protein (6xHis-NahB). Using methods of enhancing protein stability in solution, we tested different expression, cell lysis, and purification protocols with and without ligand supplementation. The protein stability was evaluated by dynamic light scattering and circular dichroism spectroscopy assays. Best-derived protocols (expression at 18 °C, cell lysis with homogenizer, and three purification steps) were used to produce 20 mg of homogeneous 6xHis-NahB per liter of culture. The secondary and quaternary structures of 6xHis-NahB were assessed by circular dichroism and size-exclusion chromatography experiments, respectively. The enzyme was NAD