MilliporeSigma
  • Differential expression of peroxiredoxin 3 in laryngeal squamous cell carcinoma.
Differential expression of peroxiredoxin 3 in laryngeal squamous cell carcinoma.
Oncotarget (2016-12-15)
Hua Zhang, Xuexia Liu, Lei Chen, Li Cai, Ning Li, Peng Zhu, Jian Chen, Xicheng Song, Guojun Li
ABSTRACT
Peroxiredoxin (PRDX) proteins are involved in carcinogenesis. PRDX3, which is predominantly localized in mitochondria and up-regulated in several human cancers, seems to confer increased treatment resistance and aggressive phenotypes. This study examined the expression profile of PRDX3 and its possible clinical value in laryngeal squamous cell carcinoma (LSCC). The expression of PRDX3 in LSCC samples was confirmed by Western blotting and further analyzed by immunohistochemistry in LSCC samples of different clinical pathological stages. The results showed that up-regulated expression of PRDX3 was observed in LSCC and associated with poor differentiation (P < 0.01), primary tumor location, N category and tumor stage (P < 0.05). Knockdown of PRDX3 in the Hep-2 laryngeal carcinoma epithelial cell line significantly enhanced Hep-2 cells' apoptosis and inhibited their proliferation and migration. Taken together, our results suggest that PRDX3 has substantial clinical impact on the progression of LSCC and shed new light on the role of PRDX3 in treatment resistance and aggressive phenotypes in LSCC.
Sigma-Aldrich
MISSION® esiRNA, targeting human PRDX3