MilliporeSigma
  • Inhibitory action of linoleamide and oleamide toward sarco/endoplasmic reticulum Ca2+-ATPase.
Inhibitory action of linoleamide and oleamide toward sarco/endoplasmic reticulum Ca2+-ATPase.
Biochimica et biophysica acta (2016-09-07)
Sachiko Yamamoto, Munenori Takehara, Makoto Ushimaru
ABSTRACT
SERCA maintains intracellular Ca2+ homeostasis by sequestering cytosolic Ca2+ into SR/ER stores. Two primary fatty acid amides (PFAAs), oleamide (18:19-cis) and linoleamide (18:29,12-cis), induce an increase in intracellular Ca2+ levels, which might be caused by their inhibition of SERCA. Three major SERCA isoforms, rSERCA1a, hSERCA2b, and hSERCA3a, were individually overexpressed in COS-1 cells, and the inhibitory action of PFAAs on Ca2+-ATPase activity of SERCA was examined. The Ca2+-ATPase activity of each SERCA was inhibited in a concentration-dependent manner strongly by linoleamide (IC50 15-53μM) and partially by oleamide (IC50 8.3-34μM). Inhibition by other PFAAs, such as stearamide (18:0) and elaidamide (18:19-trans), was hardly or slightly observed. With increasing dose, linoleamide decreased the apparent affinity for Ca2+ and the apparent maximum velocity of Ca2+-ATPase activity of all SERCAs tested. Oleamide also lowered these values for hSERCA3a. Meanwhile, oleamide uniquely reduced the apparent Ca2+ affinity of rSERCA1a and hSERCA2b: the reduction was considerably attenuated above certain concentrations of oleamide. The dissociation constants for SERCA interaction varied from 6 to 45μM in linoleamide and from 1.6 to 55μM in oleamide depending on the isoform. Linoleamide and oleamide inhibit SERCA activity in the micromolar concentration range, and in a different manner. Both amides mainly suppress SERCA activity by lowering the Ca2+ affinity of the enzyme. Our findings imply a novel role of these PFAAs as modulators of intracellular Ca2+ homeostasis via regulation of SERCA activity.
Sigma-Aldrich
cis-13-Docosenoamide
Sigma-Aldrich
Linoleamide, ≥98% (HPLC)