MilliporeSigma
  • Milk acidification to control the growth of Mycoplasma bovis and Salmonella Dublin in contaminated milk.
Milk acidification to control the growth of Mycoplasma bovis and Salmonella Dublin in contaminated milk.
Journal of dairy science (2016-10-04)
A M Parker, J K House, M S Hazelton, K L Bosward, V L Mohler, F P Maunsell, P A Sheehy
ABSTRACT
Bacterial contamination of milk fed to calves compromises calf health. Several bacterial pathogens that infect cows, including Mycoplasma bovis and Salmonella enterica ssp. enterica serovar Dublin, are shed in milk, providing a possible route of transmission to calves. Milk acidification lowers the milk pH so that it is unsuitable for bacterial growth and survival. The objectives of this study were to (1) determine the growth of M. bovis and Salmonella Dublin in milk, and (2) evaluate the efficacy of milk acidification using a commercially available acidification agent (Salstop, Impextraco, Heist-op-den-Berg, Belgium) to control M. bovis and Salmonella Dublin survival in milk. For the first objective, 3 treatments and a positive control were prepared in 10 mL of milk and broth, respectively, and inoculated with M. bovis or Salmonella Dublin to an approximate concentration of 10
Sigma-Aldrich
Deoxyribonucleic acid sodium salt from calf thymus, Type I, fibers