MilliporeSigma
  • S,S,S-Tris(2-ethylhexyl) phosphorotrithioate as an effective solvent mediator for a mexiletine-sensitive membrane electrode.
S,S,S-Tris(2-ethylhexyl) phosphorotrithioate as an effective solvent mediator for a mexiletine-sensitive membrane electrode.
Analytical and bioanalytical chemistry (2007-02-01)
Takashi Katsu, Yumi Tsunamoto, Nobumitsu Hanioka, Keiko Komagoe, Kazufumi Masuda, Shizuo Narimatsu
ABSTRACT
S,S,S-tris(2-ethylhexyl) phosphorotrithioate proved to be an effective solvent mediator for constructing a mexiletine-sensitive membrane electrode in combination with an ion-exchanger, sodium tetrakis[3,5-bis(2-methoxyhexafluoro-2-propyl)phenyl]borate. Among a series of phosphorus compounds containing phosphoryl (P=O) groups, this solvent mediator showed the highest sensitivity to mexiletine in phosphate-buffered physiological saline containing 0.15 mol L-1 NaCl and 0.01 mol L-1 NaH2PO4/Na2HPO4 (pH 7.4), giving a detection limit of 2x10(-6) mol L-1 with a slope of 58.8 mV decade-1. This is the best reported detection limit of any mexiletine-sensitive electrode developed to date. Owing to its high selectivity toward inorganic cations, the electrode was used to determine the level of mexiletine in saliva, the monitoring of which is quite effective for controlling the dose of this drug noninvasively. The mexiletine concentrations determined with the mexiletine-sensitive electrode compared favorably with those determined by high-performance liquid chromatography.