MilliporeSigma
  • Single muscle fiber proteomics reveals unexpected mitochondrial specialization.
Single muscle fiber proteomics reveals unexpected mitochondrial specialization.
EMBO reports (2015-02-04)
Marta Murgia, Nagarjuna Nagaraj, Atul S Deshmukh, Marlis Zeiler, Pasqua Cancellara, Irene Moretti, Carlo Reggiani, Stefano Schiaffino, Matthias Mann
ABSTRACT
Mammalian skeletal muscles are composed of multinucleated cells termed slow or fast fibers according to their contractile and metabolic properties. Here, we developed a high-sensitivity workflow to characterize the proteome of single fibers. Analysis of segments of the same fiber by traditional and unbiased proteomics methods yielded the same subtype assignment. We discovered novel subtype-specific features, most prominently mitochondrial specialization of fiber types in substrate utilization. The fiber type-resolved proteomes can be applied to a variety of physiological and pathological conditions and illustrate the utility of single cell type analysis for dissecting proteomic heterogeneity.
Sigma-Aldrich
Anti-IDH3A antibody produced in rabbit, IgG fraction of antiserum