MilliporeSigma
Electrochemical Behavior of Cobaltocene in Ionic Liquids.
Journal of solution chemistry (2013-03-14)
Andrzej Lewandowski, Lukasz Waligora, Maciej Galinski
ABSTRACT
The electrochemical behavior of cobaltocenium has been studied in a number of room temperature aprotic ionic liquids. Well defined, diffusion controlled, anodic and cathodic peaks were found for the Cc
Sigma-Aldrich
1-Butyl-1-methylpyrrolidinium trifluoromethanesulfonate, 95%