MilliporeSigma
  • Hyposmotic challenge modulates function of L-type calcium channel in rat ventricular myocytes through protein kinase C.
Hyposmotic challenge modulates function of L-type calcium channel in rat ventricular myocytes through protein kinase C.
Acta pharmacologica Sinica (2010-10-19)
An-tao Luo, Hong-yan Luo, Xin-wu Hu, Lin-lin Gao, Hua-min Liang, Ming Tang, Jürgen Hescheler
ABSTRACT
To study the effects and mechanisms by which hyposmotic challenge modulate function of L-type calcium current (I(Ca,L)) in rat ventricular myocytes. The whole-cell patch-clamp techniques were used to record I(Ca,L) in rat ventricular myocytes. Hyposmotic challenge(∼220 mosmol/L) induced biphasic changes of I(Ca,L), a transient increase followed by a sustained decrease. I(Ca,L) increased by 19.1%±6.1% after short exposure (within 3 min) to hyposmotic solution. On the contrary, long hyposmotic challenge (10 min) decreased I(Ca,L) to 78.1%±11.0% of control, caused the inactivation of I(Ca,L), and shifted the steady-state inactivation curve of I(Ca,L) to the right. The decreased I(Ca,L) induced by hyposmotic swelling was reversed by isoproterenol or protein kinase A (PKA) activator foskolin. Hyposmotic swelling also reduced the stimulated I(Ca,L) by isoproterenol or foskolin. PKA inhibitor H-89 abolished swelling-induced transient increase of I(Ca,L), but did not affect the swelling-induced sustained decrease of I(Ca,L). NO donor SNAP and protein kinase G (PKG) inhibitor Rp-8-Br-PET-cGMPS did not interfere with swelling-induced biphasic changes of I(Ca,L). Protein kinase C (PKC) activator PMA decreased I(Ca,L) and hyposmotic solution with PMA reverted the decreased I(Ca,L) by PMA. PKC inhibitor BIM prevented the swelling-induced biphasic changes of I(Ca,L). Hyposmotic challenge induced biphasic changes of I(Ca,L), a transient increase followed by a sustained decrease, in rat ventricular myocytes through PKC pathway, but not PKG pathway. PKA system could be responsible for the transient increase of I(Ca,L) during short exposure to hyposmotic solution.
Sigma-Aldrich
Rp-8-Bromo-β-phenyl-1,N2-ethenoguanosine 3′,5′-cyclic monophosphorothioate sodium salt, ≥98% (HPLC), powder