MilliporeSigma
  • CD146 expression correlates with epithelial-mesenchymal transition markers and a poor prognosis in gastric cancer.
CD146 expression correlates with epithelial-mesenchymal transition markers and a poor prognosis in gastric cancer.
International journal of molecular sciences (2012-07-04)
Wen-Fang Liu, Shu-Rong Ji, Jian-Jun Sun, Yi Zhang, Zhong-Yan Liu, Ai-Bin Liang, Hua-Zong Zeng
ABSTRACT
CD146 has been regarded as a novel potential therapeutic target for multiple cancers. The aim of the study was to investigate the expression of CD146 in gastric cancer and evaluate its clinical-pathological and prognostic significance. The expression of CD146 and three epithelial-mesenchymal transition (EMT)-related proteins (E-cadherin, β-catenin and vimentin) was examined in 144 gastric cancers by immunohistochemistry. Fifty-nine cases (41.0%) were defined as positive for CD146 expression. We found that CD146 expression correlated positively with lymph node involvement and a poor prognosis, and retained an independent prognostic factor for gastric cancer patients. Furthermore, positive expression of CD146 was strongly associated with loss of the epithelial marker E-cadherin and acquisition of the expression of the mesenchymal markers nuclear β-catenin and vimentin. These findings suggest that CD146 might promote EMT and progression in gastric cancer, and thus may be a potential therapeutic target for patients with gastric cancers.