MilliporeSigma
  • Anti-apoptotic effect of claudin-1 on TNF-α-induced apoptosis in human breast cancer MCF-7 cells.
Anti-apoptotic effect of claudin-1 on TNF-α-induced apoptosis in human breast cancer MCF-7 cells.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine (2012-09-04)
Yang Liu, Liang Wang, Xu-Yong Lin, Jian Wang, Juan-Han Yu, Yuan Miao, En-Hua Wang
ABSTRACT
Accumulating evidence reveals that aberrant expression of claudins manifests in various tumors; however, their biological functions are poorly understood. Here, we report on the elevated expression of claudin-1 in human breast cancer MCF-7 cells under tumor necrosis factor (TNF)-α treatment. Interestingly, the increased expression of claudin-1 contributes to an anti-apoptotic role in TNF-α-induced apoptosis. In line with this, upon TNF-α stimulus, downregulation of claudin-1 by siRNA knockdown results in a significant increase in cleavage of caspase-8 and poly (ADP-ribose) polymerase, a decrease of cyclinD1 expression, and DNA fragmentation. Consistently, TdT-mediated dUTP nick end labeling assay also shows that loss of claudin-1 increases the susceptibility of MCF-7 cells to TNF-α-induced apoptosis. However, there is no obvious effect on the expression of Bax and p53 after the treatment aforementioned. In addition, TNF-α increases the amount of claudin-1 and the cytoplasmic accumulation of β-catenin, while claudin-1 siRNA increases the amount of β-catenin in the cell membrane as well as the amount of E-cadherin in the cytoplasm. In conclusion, our data reveal a novel role of claudin-1 in regulating apoptosis in MCF-7 cells.