MilliporeSigma
  • Molecular cloning of cDNAs encoding lamp A, a human lysosomal membrane glycoprotein with apparent Mr approximately equal to 120,000.
Molecular cloning of cDNAs encoding lamp A, a human lysosomal membrane glycoprotein with apparent Mr approximately equal to 120,000.
Proceedings of the National Academy of Sciences of the United States of America (1988-06-01)
J Viitala, S R Carlsson, P D Siebert, M Fukuda
ABSTRACT
Although several lysosomal membrane glycoproteins have been characterized by using specific antibodies, none of the studies so far elucidated the amino acid sequence of a lysosomal membrane glycoprotein. Here we describe cDNA clones encoding for one of the lysosome-associated membrane proteins with apparent Mr approximately equal to 120,000, lamp A. The amino acid sequence based on the fully coded cDNA shows that as many as 18 potential N-glycosylation sites can be found in the total of 385 amino acid residues. The results obtained by endoglycosidase F digestion support the conclusion that this glycoprotein contains 18 N-glycans. These N-glycosylation sites are clustered in two domains; one contains 10 and the other contains 8 N-glycosylation sites. These domains are separated by a (proline-serine)-rich region that has a distinct homology to the IgA hinge structure. The first N-glycosylated domain is elongated to a potential leader peptide toward the NH2-terminal end. The second N-glycosylated domain, on the other hand, is connected to a putative transmembrane portion consisting of hydrophobic amino acids. This segment, in turn, is elongated to a short cytoplasmic segment composed of 11 amino acid residues at the COOH-terminal end.