MilliporeSigma
  • Inhibition of cell proliferation and migration by miR-509-3p that targets CDK2, Rac1, and PIK3C2A.
Inhibition of cell proliferation and migration by miR-509-3p that targets CDK2, Rac1, and PIK3C2A.
Molecules and cells (2014-05-08)
Sena Yoon, Eunji Han, Young-Chul Choi, Honghwan Kee, Yongsu Jeong, Jaeseung Yoon, Kwanghee Baek
ABSTRACT
CDK2 is a key regulator of cell cycle progression. In this study, we screened for miRNAs targeting CDK2 using a luciferase-3'-untranslated region reporter assay. Among 11 hit miRNAs, miR-509-3p reduced CDK2 protein levels and significantly inhibited cancer cell growth. Microarray, Western blotting, and luciferase reporter analyses revealed additional targets of miR-509-3p, including Rac1 and PIK3C2A. Overexpression of miR-509-3p induced G1 cell-cycle arrest and inhibited colony formation and migration. RNAi experiments indicated that the growth-inhibitory effects of miR-509-3p may occur through down-regulation of CDK2, Rac1, and PIK3C2A. Targeting of multiple growth regulatory genes by miR-509-3p may contribute to effective anti-cancer therapy.