MilliporeSigma
  • Localization of the tight junction protein gene TJP1 to human chromosome 15q13, distal to the Prader-Willi/Angelman region, and to mouse chromosome 7.
Localization of the tight junction protein gene TJP1 to human chromosome 15q13, distal to the Prader-Willi/Angelman region, and to mouse chromosome 7.
Genomics (1995-12-10)
T K Mohandas, X N Chen, L B Rowe, E H Birkenmeier, A S Fanning, J M Anderson, J R Korenberg
ABSTRACT
The gene encoding the tight junction (zonula occludens) protein, TJP1, was mapped to human chromosome 15q13 by fluorescence in situ hybridization (FISH) using a cDNA probe. The Jackson Laboratory backcross DNA panel derived from the cross (C57BL/6JEi x SPRET/Ei) F1 females x SPRET/Ei males was used to map the mouse Tjp1 to chromosome 7 near position 30 on the Chromosome Committee Map, a region with conserved homology to human chromosome 15q13. FISH studies on metaphases from patients with the Prader-Willi (PWS) or the Angelman syndrome (AS) showed that TJP1 maps close but distal to the PWS/AS chromosome region.