MilliporeSigma
Human syntaxin 3 is localized apically in human intestinal cells.
Journal of cell science (1997-10-24)
M H Delgrossi, L Breuza, C Mirre, P Chavrier, A Le Bivic
ABSTRACT
To understand the molecular mechanisms underlying sorting of apical and basolateral membrane components in human intestinal epithelial cells, we have cloned the human homolog of rat syntaxin 3 and looked for its subcellular localization. Endogenous human syntaxin 3 was found to be localized at the apical membrane of colon epithelial and Caco-2 cells. This apical localization was confirmed by confocal microscopy after transfection of the cDNA coding for either full length or N-terminally truncated human syntaxin 3 in Caco-2 cells. Furthermore the signal(s) and machinery targeting human syntaxin 3 to the apical membrane of epithelial cells are conserved between species since human syntaxin 3 was also localized at the apical membrane of canine MDCK cells and of epithelial cells in transgenic Drosophila melanogaster.