MilliporeSigma
  • Tuning the acceptors in catalyzed cyclizations initiated by allenes. Silylstannylation/cyclization of allene-aldehydes for synthesis of polyalkylated indolizidines including 223A congeners.
Tuning the acceptors in catalyzed cyclizations initiated by allenes. Silylstannylation/cyclization of allene-aldehydes for synthesis of polyalkylated indolizidines including 223A congeners.
The Journal of organic chemistry (2004-12-22)
Ramaiah Kumareswaran, Judith Gallucci, T V Rajanbabu
ABSTRACT
Starting from succinamide and 1,2-heptadiene-4-ol, a racemic allene-aldehyde substrate, 20, suitable for R(3)SiSnR'(3)-mediated cyclization was synthesized in six steps and in 21% yield. Stereoselective cyclization (relative cis configuration at the new stereogenic centers of the homoallyl alcohol generated) proceeded smoothly, giving a mixture of indolizidinols bearing five contiguous stereocenters in a combined yield of 80%. Relative configurations of each of the products were unequivocally established by a combination of 2D NMR experiments and single-crystal X-ray analysis. The major indolizidinol obtained in 32% yield was elaborated into indolizidine 5,8-epi-indolizidine 223A via a five-step reaction sequence in 32% overall yield. The second major component (24%) of the key cyclization yielded, in four steps, indolizidine 6,8-epi-223 in 14% yield. Even though revision of the initially postulated structure foiled our original synthetic plans for the natural product, indolizidine 223A, the new stereoselective cyclization strategy and several selective transformations of the indolizidine derivatives reported here may find further applications for the synthesis of highly alkylated indolizidine and other related alkaloids.