MilliporeSigma
  • The determination of six taste and odour compounds in water using Ambersorb 572 and high resolution mass spectrometry.
The determination of six taste and odour compounds in water using Ambersorb 572 and high resolution mass spectrometry.
The Analyst (2001-07-12)
J P Palmentier, V Y Taguchi
ABSTRACT
A method for the analysis of six taste and odour causing compounds in aqueous samples using the granular adsorbent, Ambersorb 572, and gas chromatography-high resolution mass spectrometry (GC-HRMS) has been developed. This method for the determination of geosmin, 2-methylisoborneol (2-MIB), 2-isopropyl-3-methoxypyrazine (IPMP), 2-isobutyl-3-methoxypyrazine (IBMP), 2,3,6-trichloroanisole (236-TCA) and 2,4,6-trichloroanisole (246-TCA) is highly productive [up to 40 samples per day + 23 quality control (QC) samples] and provides rapid (24-48 h) turnaround times. The analytes are extracted from water by the addition of Ambersorb 572 to the sample bottle and rolling for 1 h. The adsorbent is isolated by filtration and allowed to air dry for 1 h. The Ambersorb 572 is transferred to an autosampler vial and the analytes are desorbed into dichloromethane. The extract is analysed by GC-HRMS at 7000 resolving power (RP). Quantification is performed by isotope dilution and internal standard techniques utilizing d3-geosmin, d3-2-MIB, d5-246-TCA and 2-sec-butyl-3-methoxypyrazine (s-BMP). Method precisions of 3.5-5.8% and accuracies of +/- 5.7-8.9% were obtained. Reporting detection limits (RDLs) of 1.0 ng L-1 were obtained for 2-MIB, geosmin, IPMP and IBMP, while RDLs of 2.0 ng L-1 were obtained for 236-TCA and 246-TCA.