MilliporeSigma
  • The infrared spectra of nitriles and related compounds frozen in Ar and H2O.
The infrared spectra of nitriles and related compounds frozen in Ar and H2O.
The Astrophysical journal (1997-02-20)
M P Bernstein, S A Sandford, L J Allamandola
ABSTRACT
We present the 2320-2050 cm-1 (4.31-4.88 micrometers) infrared spectra of 16 solid-state nitriles, isonitriles, and related compounds in order to facilitate the assignment of absorption features in a spectral region now becoming accessible to astronomers for the first time through the Infrared Space Observatory (ISO). This frequency range spans the positions of the strong C triple bond N stretching vibration of these compounds and is inaccessible from the ground due to absorption by CO2 in the terrestrial atmosphere. Band positions, profiles, and intrinsic strengths (A values) were measured for compounds frozen in Ar and H2O matrices at 12 K. The molecular species examined included acetonitrile, benzonitrile (phenylcyanide), 9-anthracenecarbonitrile, dimethylcyanamide, isopropylnitrile (isobutyronitrile), methylacrylonitrile, crotononitrile, acrylonitrile (vinyl cyanide), 3-aminocrotononitrile, pyruvonitrile, dicyandiamide, cyanamide, n-butylisocyanide, methylisocyanoacetate, diisopropylcarbodiimide, and hydrogen cyanide. The C triple bond N stretching bands of the majority of nitriles fall in the 2300-2200 cm-1 (4.35-4.55 micrometers) range and have similar positions in both Ar and H2O matrices, although the bands are generally considerably broader in the H2O matrices. In contrast, the isonitriles and a few exceptional nitriles and related species produce bands at lower frequencies spanning the 2200-2080 cm-1 (4.55-4.81 micrometers) range. These features also have similar positions in both Ar and H2O matrices, and the bands are broader in the H2O matrices. Three of the compounds (pyruvonitrile, dicyandiamide, and cyanamide) show unusually large shifts of their C triple bond N stretching frequencies when changing from Ar to H2O matrices. We attribute these shifts to the formation of H2O:nitrile complexes with these compounds. The implications of these results for the identification of the 2165 cm-1 (4.62 micrometers) "XCN" interstellar feature and the 4550 cm-1 (2.2 micrometers) feature of various objects in the solar system are discussed.
Sigma-Aldrich
Pyruvonitrile, 90%