MilliporeSigma
  • Inhibition of hypoxia-inducible factor-1 function enhances the sensitivity of multiple myeloma cells to melphalan.
Inhibition of hypoxia-inducible factor-1 function enhances the sensitivity of multiple myeloma cells to melphalan.
Molecular cancer therapeutics (2009-08-13)
Yongzhen Hu, Keita Kirito, Kozue Yoshida, Toru Mitsumori, Kei Nakajima, Yumi Nozaki, Satoshi Hamanaka, Takahiro Nagashima, Masae Kunitama, Kumi Sakoe, Norio Komatsu
ABSTRACT
Abnormal activation of hypoxia-inducible factor-1 (HIF-1), one of the most important transcription factors for the adaptation of cells to hypoxia, is frequently observed in numerous types of solid tumors. Dysregulation of HIF-1 induces tumor angiogenesis and enhances the expression of anti-apoptotic proteins and glycolysis-associated enzymes in cancer cells, which in turn leads to the promotion of tumor growth. In the present study, we examined the pathophysiologic role of HIF-1 in multiple myeloma. Furthermore, we explored the possibility that HIF-1 may be a molecular target for myeloma therapy. We identified constitutive expression of the hypoxia-inducible factor-1 alpha (HIF-1alpha)-subunit in established myeloma cell lines and in primary myeloma cells. Treatment with insulin-like growth factor-1 (IGF-1) significantly increased HIF-1alpha expression through activation of the AKT and mitogen-activated protein kinase signaling pathways. Inhibition of HIF-1 function either by echinomycin, a specific HIF-1 inhibitor, or a siRNA against HIF-1alpha resulted in enhanced sensitivity to melphalan in myeloma cells. This inhibition of HIF-1 also reversed the protective effect of IGF-1 on melphalan-induced apoptosis. Inhibition of HIF-1 drastically reduced both basal and IGF-1-induced expression of survivin, one of the most important anti-apoptotic proteins in myeloma cells. We conclude that HIF-1 inhibition may be an attractive therapeutic strategy for multiple myeloma.
Sigma-Aldrich
Echinomycin, ≥98% (HPLC)