MilliporeSigma
Serum sickness-like reaction to Pamabrom.
Journal of drugs in dermatology : JDD (2006-04-01)
Jil K Swanson, Joseph C English
ABSTRACT
Pamabron is a common over-the-counter diuretic used for relief of menstrual-associated symptoms. An urticarial eruption, with systemic complaints consistent with a serum sickness-like reaction, attributed to Pamabron is described. A review of the literature concerning Pamabron and dermatology is discussed.