MilliporeSigma
  • New simple liquid chromatographic method for the determination of trimethoprim, sulfadiazine and N4-acetylsulfadiazine in plasma of broilers.
New simple liquid chromatographic method for the determination of trimethoprim, sulfadiazine and N4-acetylsulfadiazine in plasma of broilers.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2002-05-09)
G C Batzias, N A Botsoglou, V P Kotsaki-Kovatsi, G Kounenis
ABSTRACT
A new method for simultaneous quantification of trimethoprim, sulfadiazine and N4-acetylsulfadiazine in plasma of broilers at levels down to 13-16 ng/ml has been developed. Samples were deproteinized with acetonitrile, defatted with hexane, and extracted with dichloromethane. Chromatographic analysis was carried out on a C18 column in the presence of tetrabutylammonium hydrogen sulfate, a competing base, while detection was performed at 240 nm for trimethoprim, and 270 nm for both sulfadiazine and N4-acetylsulfadiazine. Accuracy and precision data showed recoveries and relative standard deviation values better than 87.3% and 3.1%, respectively, for all three analytes. The good analytical characteristics of the method could allow limits of detection in the low ng/ml range to be realised. The method was successfully applied to determine drug concentrations in plasma samples from broilers administered a combination of sulfadiazine and trimethoprim.
Product Description
Acetylsulfadiazine, European Pharmacopoeia (EP) Reference Standard