MilliporeSigma
  • Synthesis of quinazolines and tetrahydroquinazolines: copper-catalyzed tandem reactions of 2-bromobenzyl bromides with aldehydes and aqueous ammonia or amines.
Synthesis of quinazolines and tetrahydroquinazolines: copper-catalyzed tandem reactions of 2-bromobenzyl bromides with aldehydes and aqueous ammonia or amines.
Chemistry, an Asian journal (2014-01-01)
Xuesen Fan, Bin Li, Shenghai Guo, Yuanyuan Wang, Xinying Zhang
ABSTRACT
An efficient synthesis of diversely substituted quinazolines and 1,2,3,4-tetrahydroquinazolines through copper-catalyzed tandem reactions of the readily available 2-bromobenzyl bromides, aldehydes, and aqueous ammonia or amines has been developed. By using ammonia and simple aliphatic amines as the nitrogen source, the present method provides a versatile and practical protocol for the synthesis of quinazolines and 1,2,3,4-tetrahydroquinazolines.
Sigma-Aldrich
2-Bromobenzyl bromide, 98%