MilliporeSigma
  • Detection of metabolites by frequency-pulsed electron capture gas-liquid chromatography in serum and cerebrospinal fluid of a patient with Nocardia infection.
Detection of metabolites by frequency-pulsed electron capture gas-liquid chromatography in serum and cerebrospinal fluid of a patient with Nocardia infection.
Journal of clinical microbiology (1987-02-01)
J B Brooks, J V Kasin, D M Fast, M I Daneshvar
ABSTRACT
Serum (SR) and cerebrospinal fluid (CSF) from a patient suspected of having tuberculous meningitis were submitted to our laboratory for analysis by frequency-pulsed electron capture gas-liquid chromatography (FPEC GLC). The samples were tested for the presence of carboxylic acids, alcohols, hydroxy acids, and amines by methods described previously (C. C. Alley, J. B. Brooks, and D. S. Kellogg, Jr., J. Clin. Microbiol. 9:97-102, 1977; J. B. Brooks, C. C. Alley, and J. A. Liddle, Anal. Chem. 46:1930-1934, 1974; J. B. Brooks, D. S. Kellogg, Jr., M. E. Shepherd, and C. C. Alley, J. Clin. Microbiol. 11:45-51, 1980; J. B. Brooks, D. S. Kellogg, Jr., M. E. Shepherd, and C. C. Alley, J. Clin. Microbiol. 11:52-58, 1980). The results were different from previous FPEC GLC profiles of SR and CSF from patients with known tuberculous meningitis. Both the SR and CSF contained several unidentified compounds that were not previously detected in tuberculous meningitis or any of our other studies of body fluids. Nocardia brasiliensis was later isolated from the patient. Detection of these metabolites by FPEC GLC could prove to be useful for rapid diagnosis of Nocardia disease, and their identification will provide a better understanding of metabolites produced by Nocardia sp. in vivo.
Sigma-Aldrich
2-Hydroxyhexanoic acid, 98%