MilliporeSigma
  • [Analysis and assessment of meperidine and normeperidine concentrations in urine].
[Analysis and assessment of meperidine and normeperidine concentrations in urine].
Fa yi xue za zhi (1997-01-01)
M Shen, B Shen, P Xiang
ABSTRACT
In this study, urine concentrations of meperidine and normeperidine after a single therapeutic dose of meperidine in 5 healthy volunteers have been measured and compared the results to those in meperidine addicts. The results showed that there was a significant difference between two groups in the ratio of metabolite to parent drug. If can be concluded that the ratio should be aid in making a cause of meperidine injection.
Product Description
Supelco
Normeperidine solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®