MilliporeSigma
Diketene analogs as beta-lactamase inhibitor.
Chemical & pharmaceutical bulletin (1989-03-01)
K Tanizawa, K Santoh, Y Kanaoka
ABSTRACT
Compounds having a structural analogy with diketene have been synthesized and their potencies as beta-lactamase inhibitors have been studied. Among six compounds so far tested, alpha-phenyl-beta-benzylidene-3-propanolide was shown to be an irreversible inhibitor of the enzyme. The availability of simple monocyclic compounds as beta-lactamase inhibitors is discussed.