MilliporeSigma
  • Photoinduced reactions of bicycloalkylidenes with isatin and isoquinolinetrione.
Photoinduced reactions of bicycloalkylidenes with isatin and isoquinolinetrione.
Organic & biomolecular chemistry (2012-04-13)
Dong-Dong Wu, Ming-Tao He, Qi-Di Liu, Wei Wang, Jie Zhou, Lei Wang, Hoong-Kun Fun, Jian-Hua Xu, Yan Zhang
ABSTRACT
Photoinduced reactions of isatin and N-methyl-1,3,4-isoquinolinetrione with bicycloalkylidenes such as bicyclopropylidene, cyclopropylidenecyclobutane, cyclopropylidenecyclohexane and bicyclohexylidene were investigated. The reactions gave spirooxetanes as the major products derived from the [2 + 2] photocycloaddition pathway via 1,4-biradical recombination. Unusual products including the [4 + 2 + 2] cycloadducts, the oxoisochroman derivatives and other ring-rearranged products were derived from competitive pathways via 1,6-biradical recombination. The presence of oxygen in the reaction solution was found to be relevant to the distribution of different types of products. Mechanisms were proposed to rationalize the chemo- and regioselectivity in the photoreactions and the origin of the different types of products.
Sigma-Aldrich
Trimethylene oxide, 97%