MilliporeSigma
  • Structure--activity relationships of azasugar-based MMP/ADAM inhibitors.
Structure--activity relationships of azasugar-based MMP/ADAM inhibitors.
Bioorganic & medicinal chemistry letters (2003-07-23)
Hideki Moriyama, Takahiro Tsukida, Yoshimasa Inoue, Hirosato Kondo, Kohichiro Yoshino, Shin-ichiro Nishimura
ABSTRACT
In order to investigate structure-activity relationships of azasugar series toward metalloproteinases, we synthesized and evaluated several azasugar-based compounds. As a result, it was found that 4-phenoxybenzene derivative 3 having 2R,3R,4R,5S-configurations exhibited most potent inhibitory activities against matrix metalloproteinase-1, -3 and -9 and TACE.
Sigma-Aldrich
4-Phenoxybenzoic acid, 97%