MilliporeSigma
  • Release of norepinephrine from human ovary: coupling to steroidogenic response.
Release of norepinephrine from human ovary: coupling to steroidogenic response.
Endocrine (2001-11-27)
H E Lara, A Porcile, J Espinoza, C Romero, S M Luza, J Fuhrer, C Miranda, L Roblero
ABSTRACT
We investigated the possibility that norepinephrine from the human ovary is released after nerve stimulation and that this neurotransmitter is coupled to a steroidogenic response. Biologically significant levels of both norepinephrine and dopamine were found in human ovarian biopsies. [3H]norepinephrine incorporated in vitro was readily released by electrical stimulation in a Ca2+-dependent process. Ovarian membrane preparations exhibited specific binding sites for the beta-adrenergic antagonist [3H]dihydroalprenolol. Displacement of [3H]dihydroalprenolol with zinterol (a specific beta2-agonist) indicated that 72% of these sites were type beta2-receptors. beta-receptors were also present on granulosa cells. Stimulation of granulosa cells with luteinizing hormone or the beta-agonist isoproterenol increased the release of progesterone after 4 d in culture. These results suggest that the sympathetic nerves present in human ovary are coupled to beta-adrenergic receptors present in endocrine cells and, as in nonprimate mammals, appear to participate in the regulation of ovarian function.
Sigma-Aldrich
Zinterol hydrochloride, ≥98% (HPLC)