MilliporeSigma
  • Isolation and characterization of products from the nitrosation of the alkaloid gramine.
Isolation and characterization of products from the nitrosation of the alkaloid gramine.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (1985-09-01)
M U Ahmad, L M Libbey, J F Barbour, R A Scanlan
ABSTRACT
The nitrosation of gramine, a tertiary amine alkaloid present in barley malt, was carried out by reaction with sodium nitrite in buffered acetic acid (pH 3.4) for 1 hr at room temperature. Two major non-volatile products of the nitrosation reaction were isolated by preparative HPLC and characterized as indole-3-carboxylic acid and N1-nitroso-3-nitromethylindole. This interpretation was supported by spectral data. The nature of these products indicated that gramine did not undergo nitrosation by the expected mechanism of nitrosative dealkylation. A mechanism is offered to explain the labile nature of the dimethylamino group found in gramine.