Merck
Spiropyran main-chain conjugated polymers.
Macromolecular rapid communications (2012-11-22)
Michael Sommer, Hartmut Komber
ABSTRACT
The first main-chain conjugated copolymers based on alternating spiropyran (SP) and 9,9-dioctylfluorene (F8) units synthesized via Suzuki polycondensation (SPC) are presented. The reaction conditions of SPC are optimized to obtain materials of type P(para-SP-F8) with appreciably high molecular weights up to M(w) ≈ 100 kg mol(-1). (13)C NMR is used to identify the random orientation of the non-symmetric SP unit in P(p-SP-F8). Ultrasound-induced isomerization of P(p-SP-F8) to the corresponding merocyanine form P(p-MC-F8) yields a deep-red solution. This isomerization reaction is followed by (1)H NMR in solution using sonication, whereby the color increasingly changes to deep red. The possibility to incorporate multiple SP units into main-chain polymers significantly broadens existing SP-based polymeric architectures.
Product Description
Supelco
Fluorene, analytical standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5000 mg
Please contact Customer Service for Availability
65,10 €
Sigma-Aldrich
Fluorene, 98%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
Please contact Customer Service for Availability
26,50 €
100 g
Please contact Customer Service for Availability
75,70 €
500 g
Please contact Customer Service for Availability
198,00 €