MilliporeSigma
  • In situ absorption and relative bioavailability studies of zaleplon loaded self-nanoemulsifying powders.
In situ absorption and relative bioavailability studies of zaleplon loaded self-nanoemulsifying powders.
Journal of microencapsulation (2012-08-17)
Karthik Y Janga, Raju Jukanti, Sharath Sunkavalli, Ashok Velpula, Suresh Bandari, Prabhakar Kandadi, Prabhakar Reddy Veerareddy
ABSTRACT
Self-nanoemulsifying drug delivery systems (SNEDDSs) offer potential as suitable carriers for improved oral delivery of poorly soluble and low bioavailable drugs. To derive self-nanoemulsifying powders (SNEPs), the optimized Z-SNEDDS formulation was adsorbed onto different carriers and based on micromeritics the formulation loaded onto neusilin US2 (SNEP-N) was selected for further characterization. The solid-state characterization (scanning electron microscopy, differential scanning calorimetry and powder X-ray diffraction) studies unravel the transformation of native crystalline state to amorphous and/or molecular state. The higher predictive effective permeability coefficient and fraction absorbed in humans extrapolated from in situ single-pass intestinal absorption study data in rats provide an insight on the potential of SNEPs for augment in absorption across gastrointestinal barrier. Overall a 3.5-fold enhancement in the extent of absorption of zaleplon from SNEP-N formulation proves the feasibility of SNEPs formulation for improved oral delivery of zaleplon.
Product Description
Supelco
Zaleplon solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®