MilliporeSigma
  • Botanical insecticides in controlling Kelly's citrus thrips (Thysanoptera: Thripidae) on organic grapefruits.
Botanical insecticides in controlling Kelly's citrus thrips (Thysanoptera: Thripidae) on organic grapefruits.
Journal of economic entomology (2012-02-04)
V A Vassiliou
ABSTRACT
Kelly's citrus thrips, Pezothrips kellyanus (Bagnall) (Thysanoptera: Thripidae) was first recorded in Cyprus in 1996 and became an economic citrus pest. In Cyprus, Kelly's citrus thrips larvae cause feeding damage mainly on immature lemon and grapefruit fruits. Use of botanical insecticides is considered an alternative tool compared with synthetic chemicals, in offering solutions for healthy and sustainable citrus production. During 2008-2010, the efficacy of the botanical insecticides azadirachtin (Neemex 0.3%W/W and Oikos 10 EC), garlic extract (Alsa), and pyrethrins (Vioryl 5%SC) was evaluated in field trials against Kelly's citrus thrips larval stage I and II aiming at controlling the pest's population and damage to organic grapefruit fruits. In each of the trial years treatments with pyrethrins and azadirachtin (Neemex 0.3%W/W) were the most effective against Kelly's citrus thrips compared with the untreated control (for 2008: P < 0.018; for 2009: P < 0.000; for 2010: P < 0.008). In 2008, the mean number of damaged fruits in treatments with pyrethrins and Neemex was 9.6 (19.2%) and 9.7 (19.5%) respectively, compared with 12.2 (24.3%) in the untreated control. In 2009, the mean number of damaged fruits in treatment with pyrethrins was 3.7 (7.3%) and 3.9 (7.8%) in treatment with Neemex compared with 8.6 (17.3%) in the untreated control, while in 2010 the mean damaged fruits in these treatments was recorded at 18.7 (37.5%) and 19.6 (39.2), respectively, compared with 29.6 fruits (59.2%) in the control. Oikos 10 EC showed significant effect only in 2009 and 2010. In these years, the mean number of damaged fruits was recorded at 5.5 and 21.2 compared with 8.6 and 29.6 fruits in the untreated control, respectively. Garlic extract showed the lowest effect from all the botanicals used compared with the untreated control.
Sigma-Aldrich
Azadirachtin, ~95%