MilliporeSigma
  • Metabolism of methylisoeugenol in liver microsomes of human, rat, and bovine origin.
Metabolism of methylisoeugenol in liver microsomes of human, rat, and bovine origin.
Drug metabolism and disposition: the biological fate of chemicals (2011-06-03)
Alexander T Cartus, Karl-Heinz Merz, Dieter Schrenk
ABSTRACT
Methylisoeugenol (1,2-dimethoxy-4-propenylbenzene, 1) is a minor constituent of essential oils, naturally occurring as a mixture of cis/trans isomers. 1 is a U.S. Food and Drug Administration-approved food additive and has been given "Generally Recognized as Safe" status. Previously, metabolism of 1 has been studied in the rat, revealing mainly nontoxic cinnamoyl derivatives as major metabolites. However, data concerning the possible formation of reactive intermediary metabolites are not available to date. In this study, the oxidative metabolism of 1 was studied using liver microsomes of rat [not induced, rat liver microsomes (RLM); Aroclor1254 induced RLM (ARLM)], bovine, and human (pooled from 150 donors) origin. Incubations of these microsomes with 1 provided phase I metabolites that were separated by high-performance liquid chromatography (HPLC) and identified by NMR and UV-visible spectroscopy and/or liquid chromatography-mass spectrometry. Identity was confirmed by comparison with (1)H NMR spectra of synthesized reference compounds. Formation of metabolites was quantified by HPLC/UV using dihydromethyleugenol (10) synthesized as the internal standard. From incubations of ARLM with 1, seven metabolites could be detected, with 3'-hydroxymethylisoeugenol (2), isoeugenol and isochavibetol (3 + 4), and 6-hydroxymethylisoeugenol (5) being the main metabolites. Secondary metabolites derived from 1 were identified as the α,β-unsaturated aldehyde 3'-oxomethylisoeugenol (6) and 1',2'-dihydroxy-dihydromethylisoeugenol (7). We were surprised to find that formation of allylic 6-hydroxymethyleugenol (8) was observed starting at approximately 30 min after the beginning of incubations with ARLM. HLM did not form ring-hydroxylated metabolites but were most active in the formation of 6 and 7. ARLM incubations displayed the highest turnover rate and broadest metabolic pattern, presumably resulting from an increased expression of cytochrome P450 enzymes. In conclusion, we present a virtually complete pattern of nonconjugated microsomal metabolites of 1 comprising reactive metabolites and suggest the formation of reactive intermediates that need more investigation with respect to their possible adverse properties.
Sigma-Aldrich
1,2-Dimethoxy-4-propenylbenzene, 99%
Sigma-Aldrich
Methyl isoeugenol, ≥98%, FG