MilliporeSigma
  • Do corticosteroids affect lumbar spinal fusion? A rabbit model using high-dose methylprednisolone.
Do corticosteroids affect lumbar spinal fusion? A rabbit model using high-dose methylprednisolone.
Journal of orthopaedic science : official journal of the Japanese Orthopaedic Association (2011-04-30)
Julio Urrutia, Maximiliano Carmona, Jorge Briceno
ABSTRACT
The effect of corticosteroids on spinal fusion healing has not yet been determined. To evaluate the effect of corticosteroids on lumbar spinal fusion we designed a randomized, placebo-controlled animal study using high-dose methylprednisolone sodium succinate, which is widely used in patients with spinal cord injury who are undergoing spinal fusion. Two groups of 18 rabbits underwent a postero-lateral fusion at L5-L6 with autologous bone graft. After surgery, the animals were assigned to receive: (a) methylprednisolone sodium succinate 30 mg/kg over 15 min, followed by an intravenous infusion of 5.4 mg/kg/h for 23 h, or (b) normal saline in the same volume. Animals were killed 8 weeks after surgery; the presence of fusion was analyzed by use of two different methods: a manual palpation test and an antero-posterior radiograph. Both groups of animals were comparable in weight. Fusion was achieved in 5/18 rabbits (27.8%) in the methylprednisolone group and in 9/18 animals (50%) in the control group (p = 0.17). In a lumbar posterolateral fusion rabbit model, high-dose methylprednisolone sodium succinate reduced the success of lumbar fusion; however, our data did not reach statistical significance.
Sigma-Aldrich
6α-Methylprednisolone 21-hemisuccinate sodium salt, lyophilized powder