MilliporeSigma
  • Development of a real-time RT-PCR assay combined with ethidium monoazide treatment for RNA viruses and its application to detect viral RNA after heat exposure.
Development of a real-time RT-PCR assay combined with ethidium monoazide treatment for RNA viruses and its application to detect viral RNA after heat exposure.
Water science and technology : a journal of the International Association on Water Pollution Research (2011-02-01)
K Kim, H Katayama, M Kitajima, Y Tohya, S Ohgaki
ABSTRACT
A method was developed for discriminating damaged viruses or naked viral RNA from intact viruses by ethidium monoazide (EMA) treatment before RT-PCR. The applied EMA treatment consisted of three steps: (1) EMA dose, (2) exposure to light, and (3) additional purification by spin-column gel filtration. Approximately 4-log reduction in viral RNA concentration was observed by adding a dose of 10 μg/mL-EMA with 300 s of light irradiation. Although residual EMA can be an inhibitor of RT-PCR, its effect was reduced by spin-column gel filtration or a QIAamp® Viral RNA Mini Kit. EMA-RT-PCR was applied to the thermally treated PV1. Results of EMA-RT-PCR were similar to the plaque assay when PV1 was thermally inactivated. Although this is a preliminary study investigating applicability of the EMA-RT-PCR method for RNA viruses, the results suggest that the method is potentially applicable for the selective detection of epidemiologically important enteric viruses in water such as enteroviruses and noroviruses.
Sigma-Aldrich
Ethidium bromide monoazide, ≥95% (HPLC), solid