MilliporeSigma
  • [Determination of hormone multi-residues in animal tissues by gas chromatography-mass spectrometry].
[Determination of hormone multi-residues in animal tissues by gas chromatography-mass spectrometry].
Se pu = Chinese journal of chromatography (2009-10-07)
Weixuan Lin, Weifeng Dong, Xi Chen, Miao Tian, Ling Yu, Jinghong Zhao, Chunguang Yang
ABSTRACT
A method of gas chromatography-mass spectrometry (GC-MS) for the simultaneous determination of nine sex hormone residues, such as hexestrol, diethylstilbestrol, dienestrol, etiocholan-3alpha-ol-17-one, epitestosterone, estrone, estradiol, ethinylestradiol and estriol, in animal tissues was developed. The sex hormones were extracted with acetonitrile, then cleaned-up with a C18 solid-phase extraction (SPE) column. The microwave-assisted derivatization of the target components with N,O-bis( trimethylsilyl) trifluoroacetamide (BSTFA) and trimethylchlorosilane (TMCS) (99:1, v/v) using pyridine as solvent was performed, and then the derivatives were analyzed by GC-MS. The limits of detection were 0.1-1 microg/kg for all hormones, and the limits of quantification were 0.2-2 microg/kg. The average recoveries of sex hormones were 68.8%-93.1%. The relative standard deviations (RSDs) of inter- and intra-assay were 4.1%-22.3% and 3.1%-17.9%, respectively. The real sample tests showed this method can be used for the sensitive and accurate determination of multi-sex hormones residues in biological samples.