MilliporeSigma
  • Rational design and synthesis of potent dibenzazepine motifs as beta-secretase inhibitors.
Rational design and synthesis of potent dibenzazepine motifs as beta-secretase inhibitors.
Journal of medicinal chemistry (2009-10-01)
Taleb H Al-Tel, Raed A Al-Qawasmeh, Marco F Schmidt, Amal Al-Aboudi, Shashidhar N Rao, Salim S Sabri, Wolfgang Voelter
ABSTRACT
We have identified small-molecule dibenzazepine inhibitors of beta-secretase (BACE1). These BACE1 inhibitors possess two key salient features. The first is a seven-membered heterocyclic ring fused to two aromatic rings representing the P3-P2 residues. The second is an amide and/or amide bioisostere representing the P1' residue. Rational optimization led to the identification of potent analogues, such as 10 (K(I) = 211 nM).
Sigma-Aldrich
Isophthalic acid, 99%