MilliporeSigma
  • [Mechanisms of action of plant resistance inductor beta-aminobutyric acid in Allium cepa].
[Mechanisms of action of plant resistance inductor beta-aminobutyric acid in Allium cepa].
TSitologiia i genetika (2009-03-04)
S A Poliakovskiĭ, Zh N Kravchuk, A P Dmitriev
ABSTRACT
The effects of beta-aminobutiric acid as an inductor of plant resistance was studied. The influence of BABA pretreatment on A. cepa plants of three different pathogen-resistant varieties was investigated by fluorescence microscopic methods. It was determined that BABA is able to prime the plant callose accumulation.
Sigma-Aldrich
3-Aminobutanoic acid, 97%
Sigma-Aldrich
(S)-3-Aminobutyric acid, 97%