MilliporeSigma
  • Mosquito larvicidal activity of active constituent derived from Chamaecyparis obtusa leaves against 3 mosquito species.
Mosquito larvicidal activity of active constituent derived from Chamaecyparis obtusa leaves against 3 mosquito species.
Journal of the American Mosquito Control Association (2006-03-02)
Young-Su Jang, Ju-Hyun Jeon, Hoi-Seon Lee
ABSTRACT
Mosqutio larvicidal activity of Chamaecyparis obtusa leaf-derived materials against the 4th-stage larvae of Aedes aegypti (L.), Ochlerotatus togoi (Theobald), and Culex pipiens pallens (Coquillett) was examined in the laboratory. A crude methanol extract of C. obtusa leaves was found to be active (percent mortality rough) against the 3 species larvae; the hexane fraction of the methanol extract showed a strong larvicidal activity (100% mortality) at 100 ppm. The bioactive component in the C. obtusa leaf extract was characterized as beta-thujaplicin by spectroscopic analyses. The LC50 value of beta-thujaplicin was 2.91, 2.60, and 1.33 ppm against Ae. aegypti, Oc. togoi, and Cx. pipiens pallens larvae. This naturally occurring C. obtusa leaves-derived compound merits further study as a potential mosquito larval control agent or lead compound.
Product Description
Supelco
Pirimiphos-methyl, PESTANAL®, analytical standard