MilliporeSigma
  • Stable planar six-pi-electron six-membered N-heterocyclic carbenes with tunable electronic properties.
Stable planar six-pi-electron six-membered N-heterocyclic carbenes with tunable electronic properties.
Journal of the American Chemical Society (2005-07-21)
Carsten Präsang, Bruno Donnadieu, Guy Bertrand
ABSTRACT
Carbene analogues of borazines are highly thermally stable. Keeping quasi-identical steric demands, the electronic properties of the carbene can be precisely tuned by varying the nature of the substituents at the boron centers.
Sigma-Aldrich
Formamidine acetate salt, 99%
Sigma-Aldrich
Formamidine hydrochloride, 97%