MilliporeSigma
  • Cyanidin and Malvidin from Oryza sativa cv. Heugjinjubyeo mediate cytotoxicity against human monocytic leukemia cells by arrest of G(2)/M phase and induction of apoptosis.
Cyanidin and Malvidin from Oryza sativa cv. Heugjinjubyeo mediate cytotoxicity against human monocytic leukemia cells by arrest of G(2)/M phase and induction of apoptosis.
Journal of agricultural and food chemistry (2004-04-15)
Jin Won Hyun, Ha Sook Chung
ABSTRACT
Oryza sativa cv. Heugjinjubyeo (Gramineae), anthocyanin-pigmented rice, having dark purple grains, is known broadly as enriched rice with an improved taste. Two bioactive compounds were isolated from the 0.5% HCl-ethyl alcohol soluble fraction of the aleurone layer of O. sativa cv. Heugjinjubyeo through an activity-monitored fractionation and isolation method. From spectral analysis, the cytotoxic components were the anthocyanidins cyanidin (1) and malvidin (2) The 50% growth inhibitory concentrations (IC(50)) of cyanidin and malvidin on U937, human monocytic leukemia cells, were 60 and 40 microg/mL, respectively. These compounds showed cytotoxicity through the arrest of the G(2)/M phase of cell cycle and induction of apoptosis.
Sigma-Aldrich
Malvidin chloride, ≥95.0% (HPLC)