MilliporeSigma
  • Some 1,2-diphenylethane derivatives as inhibitors of retinoic acid--metabolising enzymes.
Some 1,2-diphenylethane derivatives as inhibitors of retinoic acid--metabolising enzymes.
Journal of enzyme inhibition and medicinal chemistry (2003-12-25)
V P Greer, P Mason, A J Kirby, H J Smith, P J Nicholls, C Simons
ABSTRACT
In a search for novel inhibitors of RA-metabolising enzyme inhibitors as potential anti-cancer agents some 1,2-ethandiones, 2-hydroxyethanones and 1-ethylenedioxyethanones based on aryl-substituted 1,2-diphenylethane have been examined. Several of the compounds were weak inhibitors of the non-specific rat liver microsomal P450 enzymes and moderate inhibitors of the RA-induced enzymes in cultured human genital fibroblasts, where the RA-specific enzyme CYP26 is probably expressed. The 2-hydroxyethanone (13) with a 1-(4-dimethylaminophenyl) substituent was overall the most potent compound for rat liver microsomal enzyme (IC50 = 52.1 microM; ketoconazole, 2.8 microM) and the RA-induced enzyme (100 microM, 65.9% inhibition; ketoconazole, 20 microM, 75.0%). Modification of the dimethylamino group in (13) with more hydrophobic dialkylamino functions or separate modification of the 2-(2,4-dichlorophenyl) function did not improve potency.
Sigma-Aldrich
Bibenzyl, ReagentPlus®, 99%