MilliporeSigma
  • Three-dimensional pharmacophore hypotheses for the locust neuronal octopamine receptor (OAR3). Part 2: agonists.
Three-dimensional pharmacophore hypotheses for the locust neuronal octopamine receptor (OAR3). Part 2: agonists.
Bioorganic & medicinal chemistry (1999-08-28)
A Hirashima, C Pan, E Kuwano, E Taniguchi, M Eto
ABSTRACT
Three-dimensional pharmacophore hypotheses were built from a set of 43 agonists against octopamine receptor class 3 (OAR3) in locust nervous tissue. Among the 10 chemical-featured models generated by program Catalyst/Hypo, a hypothesis including hydrogen-bond acceptor (HBA), hydrophobic (Hp), and hydrophobic aliphatic (HpA1) features was considered to be important and predictive in evaluating OAR3 agonists. While the ideal and null hypotheses had a cost of 156.40 and 239.20, respectively, the 10 resulting hypotheses possessed costs from 169.89 to 175.81. The best hypothesis that was confirmed to have a 95% chance of true correlation yielded a low RMS of 0.757 and high regression r of 0.933. Active agonists mapped well onto all the features of the hypothesis such as HBA, Hp, and HpA1. On the other hand, inactive compounds were shown to be difficult to achieve the energetically favorable conformation which is found in the active molecules in order to fit the 3-D chemical feature pharmacophore models.
Sigma-Aldrich
1-Phenylimidazole, 97%