MilliporeSigma
  • ARNTL2 upregulation of ACOT7 promotes NSCLC cell proliferation through inhibition of apoptosis and ferroptosis.
ARNTL2 upregulation of ACOT7 promotes NSCLC cell proliferation through inhibition of apoptosis and ferroptosis.
BMC molecular and cell biology (2023-04-03)
Tao Wang, Kai Wang, Xu Zhu, Nan Chen
ABSTRACT
Recent studies have reported that the circadian transcription factor aryl hydrocarbon receptor nuclear translocator like 2 (ARNTL2) promotes the metastatic progression of lung adenocarcinoma. However, the molecular mechanisms of ARNTL2 in non-small cell lung cancer (NSCLC) cell growth and proliferation remain to be explored. The expression of ARNTL2 and acyl-CoA thioesterase 7 (ACOT7) in lung cancer patients was analyzed based on TCGA database. Gain-of-function of ARNTL2 and ACOT7 was conducted by transfecting the cells with plasmids or lentivirus. Knockdown assay was carried out by siRNAs. Western blot and qRT-PCR were performed to check the protein and mRNA expression. Dual luciferase and ChIP-qPCR assay was applied to check the interaction of ARNTL2 on ACOT7's promoter sequence. Triglyceride level, MDA production, the activity of casapase 3 to caspase 7, and lipid ROS were measured by indicated assay kit. Cellular function was detected by CCK8, colony formation and flow cytometry analysis of cell death and cell cycle. We demonstrated that ARNTL2 upregulation of ACOT7 was critical for NSCLC cell growth and proliferation. Firstly, overexpression of ARNTL2 conferred the poor prognosis of LUAD patients and supported the proliferation of NSCLC cells. Based on molecular experiments, we showed that ARNTL2 potentiated the transcription activity of ACOT7 gene via direct binding to ACOT7's promoter sequence. ACOT7 high expression was correlated with the worse prognosis of LUAD patients. Gain-of-function and loss-of-function experiments revealed that AOCT7 contributed to NSCLC cell growth and proliferation. ACOT7 regulated the apoptosis and ferroptosis of NSCLC cells, while exhibited no effect on cell cycle progression. ACOT7 overexpression also potentiated fatty acid synthesis and suppressed lipid peroxidation. Lastly, we showed that ARNTL2 knockdown and overexpression inhibited and promoted the cellular triglyceride production and subsequent cell proliferation, which could be reversed by ACOT7 overexpression and knockdown. Our study illustrated the oncogenic function of ARNTL2/ACOT7 axis in the development of NSCLC. Targeting ARNTL2 or ACOT7 might be promising therapeutic strategies for NSCLC patients with highly expressed ARNTL2.
Sigma-Aldrich
Anti-ARNTL2 antibody produced in rabbit, affinity isolated antibody