MilliporeSigma
  • Effect of human adipose tissue-derived mesenchymal-stem-cell bioactive materials on porcine embryo development.
Effect of human adipose tissue-derived mesenchymal-stem-cell bioactive materials on porcine embryo development.
Molecular reproduction and development (2013-10-24)
Hyo-Young Park, Eun-Young Kim, Seung-Eun Lee, Hyun-Yong Choi, Jeremiah Jiman Moon, Min-Jee Park, Yeo-Jin Son, Jun-Beom Lee, Chang-Jin Jeong, Dong-Sun Lee, Key-Jung Riu, Se-Pill Park
ABSTRACT
Human adipose tissue-derived mesenchymal stem cells (hAT-MSCs) secrete bioactive materials that are beneficial for tissue repair and regeneration. In this study, we characterized human hAT-MSC bioactive material (hAT-MSC-BM), and examined the effect of hAT-MSC-BM on porcine embryo development. hAT-MSC-BM was enriched with several growth factors and cytokines, including fibroblast growth factor 2 (FGF2), vascular endothelial growth factor A (VEGFA), and interleukin 6 (IL6). Among the various concentrations and days of treatment tested, 10% hAT-MSC-BM treatment beginning on culture Day 4 provided the best environment for the in vitro growth of parthenogenetic porcine embryos. While the addition of 10% fetal bovine serum (FBS) increased the hatching rate and the total cell number of parthenogenetic porcine embryos compared with the control and hAT-MSC culture medium group, the best results were from the group cultured with 10% hAT-MSC-BM. Mitochondrial activity was also higher in the 10% hAT-MSC-BM-treated group. Moreover, the relative mRNA expression levels of development and anti-apoptosis genes were significantly higher in the 10% hAT-MSC-BM-treated group than in control, hAT-MSC culture medium, or 10% FBS groups, whereas the transcript abundance of an apoptosis gene was slightly lower. Treatment with 10% hAT-MSC-BM starting on Day 4 also improved the development rate and the total cell number of in vitro-fertilized embryos. This is the first report on the benefits of hAT-MSC-BM in a porcine embryo in vitro culture system. We conclude that hAT-MSC-BM is a new, alternative supplement that can improve the development of porcine embryos during both parthenogenesis and fertilization in vitro.
Sigma-Aldrich
Anti-CD34 Class I Antibody, clone B1-3C5, clone B1-3C5, Chemicon®, from mouse