MilliporeSigma
  • Insulinoma-associated protein 1 immunostaining for various types of neuroendocrine tumors on FNA smears.
Insulinoma-associated protein 1 immunostaining for various types of neuroendocrine tumors on FNA smears.
Cancer cytopathology (2020-06-24)
Tieying Hou, Qiong Gan, Cicily T Joseph, Xiaoping Sun, Yun Gong
ABSTRACT
Insulinoma-associated protein 1 (INSM1) has recently emerged as a reliable nuclear immunostaining marker for detecting neuroendocrine tumors (NETs) in paraffin-embedded surgical samples and cytologic cell blocks, but the reliability of INSM1 staining on cytologic smears is understudied. This study investigated the performance of INSM1 staining on cytologic smears for the detection of various NETs in comparison with chromogranin (CG) and synaptophysin (SYN). INSM1, CG, and SYN were stained on cytologic smears of 70 NETs, including 20 pancreatic NETs, 10 lung carcinoid tumors, 11 small cell lung carcinomas (SCLCs), 10 medullary thyroid carcinomas, 10 Merkel cell carcinomas, 4 thymic atypical carcinoid tumors, and 5 olfactory neuroblastomas. The detection rate, the percentage of positive cells, and the staining intensity were recorded. The overall detection rate of INSM1 (94%) was higher than the rates of CG (79%) and SYN (89%). The detection rate of INSM1 was higher than the rates of CG and SYN in SCLC, Merkel cell carcinoma, and olfactory neuroblastoma; higher than the rate of CG and equal to the rate of SYN in pancreatic NETs and medullary thyroid carcinoma; equal to the rate of CG and higher than the rate of SYN in thymic atypical carcinoid tumors; and equal to the rate of CG and lower than the rate of SYN in lung carcinoid tumors. INSM1 staining was easier to interpret than CG and SYN staining, especially in high-grade NETs. INSM1 can be reliably stained on cytologic smears and outperforms CG and SYN in the verification of clinically or radiologically suspected NETs.
Sigma-Aldrich
Anti-Chromogranin A Antibody, clone LK2H10, clone LK2H10, Chemicon®, from mouse