MilliporeSigma
  • Effects of Titanium Dioxide Nanoparticles on Red Clover and Its Rhizobial Symbiont.
Effects of Titanium Dioxide Nanoparticles on Red Clover and Its Rhizobial Symbiont.
PloS one (2016-05-14)
Janine Moll, Annette Okupnik, Alexander Gogos, Katja Knauer, Thomas D Bucheli, Marcel G A van der Heijden, Franco Widmer
ABSTRACT
Titanium dioxide nanoparticles (TiO2 NPs) are in consideration to be used in plant protection products. Before these products can be placed on the market, ecotoxicological tests have to be performed. In this study, the nitrogen fixing bacterium Rhizobium trifolii and red clover were exposed to two TiO2 NPs, i.e., P25, E171 and a non-nanomaterial TiO2. Growth of both organisms individually and their symbiotic root nodulation were investigated in liquid and hydroponic systems. While 23 and 18 mg l(-1) of E171 and non-nanomaterial TiO2 decreased the growth rate of R. trifolii by 43 and 23% respectively, P25 did not cause effects. Shoot length of red clover decreased between 41 and 62% for all tested TiO2 NPs. In 21% of the TiO2 NP treated plants, no nodules were found. At high concentrations certain TiO2 NPs impaired R. trifolii as well as red clover growth and their symbiosis in the hydroponic systems.
Sigma-Aldrich
Titanium(IV) oxide, anatase, powder, 99.8% trace metals basis