MilliporeSigma
  • Bystander activation of CD8+ T cells contributes to the rapid production of IFN-gamma in response to bacterial pathogens.
Bystander activation of CD8+ T cells contributes to the rapid production of IFN-gamma in response to bacterial pathogens.
Journal of immunology (Baltimore, Md. : 1950) (2001-01-06)
G Lertmemongkolchai, G Cai, C A Hunter, G J Bancroft
ABSTRACT
The bacterium Burkholderia pseudomallei causes a life-threatening disease called melioidosis. In vivo experiments in mice have identified that a rapid IFN-gamma response is essential for host survival. To identify the cellular sources of IFN-gamma, spleen cells from uninfected mice were stimulated with B. pseudomallei in vitro and assayed by ELISA and flow cytometry. Costaining for intracellular IFN-gamma vs cell surface markers demonstrated that NK cells and, more surprisingly, CD8(+) T cells were the dominant sources of IFN-gamma. IFN-gamma(+) NK cells were detectable after 5 h and IFN-gamma(+) CD8(+) T cells within 15 h after addition of bacteria. IFN-gamma production by both cell populations was inhibited by coincubation with neutralizing mAb to IL-12 or IL-18, while a mAb to TNF had much less effect. Three-color flow cytometry showed that IFN-gamma-producing CD8(+) T cells were of the CD44(high) phenotype. The preferential activation of NK cells and CD8(+) T cells, rather than CD4(+) T cells, was also observed in response to Listeria monocytogenes or a combination of IL-12 and IL-18 both in vitro and in vivo. This rapid mechanism of CD8(+) T cell activation may be an important component of innate immunity to intracellular pathogens.
Sigma-Aldrich
Anti-Cd8a low endotoxin antibody, Rat monoclonal, clone 53-6.7, purified immunoglobulin, buffered aqueous solution