MilliporeSigma
  • Production of 10-hydroxystearic acid from oleic acid and olive oil hydrolyzate by an oleate hydratase from Lysinibacillus fusiformis.
Production of 10-hydroxystearic acid from oleic acid and olive oil hydrolyzate by an oleate hydratase from Lysinibacillus fusiformis.
Applied microbiology and biotechnology (2011-12-23)
Bi-Na Kim, Young-Chul Joo, Yeong-Su Kim, Kyoung-Rok Kim, Deok-Kun Oh
ABSTRACT
A recombinant enzyme from Lysinibacillus fusiformis was expressed, purified, and identified as an oleate hydratase because the hydration activity of the enzyme was the highest for oleic acid (with a k (cat) of 850 min(-1) and a K (m) of 540 μM), followed by palmitoleic acid, γ-linolenic acid, linoleic acid, myristoleic acid, and α-linolenic acid. The optimal reaction conditions for the enzymatic production of 10-hydroxystearic acid were pH 6.5, 35 °C, 4% (v/v) ethanol, 2,500 U ml(-1) (8.3 mg ml(-1)) of enzyme, and 40 g l(-1) oleic acid. Under these conditions, 40 g l(-1) (142 mM) oleic acid was converted into 40 g l(-1) (133 mM) 10-hydroxystearic acid for 150 min, with a molar yield of 94% and a productivity of 16 g l(-1) h(-1), and olive oil hydrolyzate containing 40 g l(-1) oleic acid was converted into 40 g l(-1) 10-hydroxystearic acid for 300 min, with a productivity of 8 g l(-1) h(-1).
Sigma-Aldrich
Myristoleic acid, ≥99% (capillary GC)