MilliporeSigma
  • Analysis of wax ester molecular species by high performance liquid chromatography/atmospheric pressure chemical ionisation mass spectrometry.
Analysis of wax ester molecular species by high performance liquid chromatography/atmospheric pressure chemical ionisation mass spectrometry.
Journal of chromatography. A (2010-01-19)
Vladimír Vrkoslav, Klára Urbanová, Josef Cvacka
ABSTRACT
High chromatographic resolution of wax esters (WEs) was achieved by non-aqueous reversed-phase liquid chromatography on a Nova-Pak C18 column by optimising the acetonitrile/ethyl acetate mobile phase gradient. The retention behaviour of WEs was studied in this chromatographic system. The WEs eluted according to their equivalent carbon number (ECN) values; within the group of WEs with the identical ECN, the most unsaturated species tended to elute first. The isobaric WEs with different positions of the ester moiety were separated from each other whenever the lengths of the chains were sufficiently different. The methyl-branched esters eluted at shorter retention times than the straight-chained analogues, and the resolution among methyl-branched WEs depended on the position of the branching. The analytes were detected by atmospheric pressure chemical ionisation mass spectrometry (APCI-MS) using data-dependent scanning. WEs provided simple full-scan spectra with abundant protonated molecules and low-intensity fragments. Collision-induced dissociation (CID) promoted identification of the WE molecular species. The responses of WEs were found to be dependent on the number of double bonds and on the alkyl-chain length; the limits of the detection ranged from 20micromol/L to 200nmol/L. The HPLC/APCI-MS was applied for the analysis of the WEs isolated from honeycomb beeswax, jojoba oil and human hair. Good agreement between reported results and the literature data was achieved, with several novel polyunsaturated WEs also being found.